missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
£30.10 - £54.85
Specifications
Format | Strip |
---|---|
For Use With (Application) | Routine pH testing |
Dimensions (L x W) | 100 x 11 mm |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
10451602
|
Cytiva
2627-990 |
3.8 to 5.5 |
£54.85
Pack of 200 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11370684
|
Cytiva
2612-990 |
1 to 12 |
£46.18
Pack of 200 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10114071
|
Cytiva
2626-990 |
1.8 to 3.8 |
£51.40
Pack of 200 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
10614282
|
Cytiva
2628-990 |
5.2 to 6.8 |
£30.10
Pack of 200 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11330694
|
Cytiva
2629-990 |
6 to 8.1 |
£51.80
Pack of 200 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11340694
|
Cytiva
2630-990 |
8 to 9.7 |
£33.25
Pack of 200 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
11914556
|
Cytiva
2631-990 |
9.5 to 12 |
£34.10
Pack of 200 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Strip | |
Routine pH testing | |
100 x 11 mm |