Learn More
Cytiva Whatman™ pH Indicators Reel Dispensers
Used for rapid determination of pH values
Brand: Cytiva 2600-102A
This item is not returnable.
View return policy
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
4 to 7 | |
Reel | |
Routine pH testing | |
pH Indicator Dispenser |
Product Content Correction
Product Title
Cytiva Whatman™ pH Indicators Reel Dispensers > pH 4-7 range; 1/Box
Spot an opportunity for improvement?Share a Content Correction