missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap Disposable Capsules, 150 TF

Polycap 150 TF filters for filtration of organic solutions, aggressive solutions, air and gas

Brand:  Cytiva 2802T

 View more versions of this product

Product Code. 11359164

  • £682.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Integrity testable
  • Bidirectional flow: certain applications may require orientation
  • Filter Media: Hydrophobic PTFE membrane

Applications:

Filtration of organic solutions, aggressive solutions, air and gas; also can be used as vent.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
PTFE
Polypropylene (PP)
4.1 bar (60 psi)
Polycap TF 150, 0.2 μm, with FNPT inlet and outlet (5 pcs)
0.2 μm
Female national pipe thread (FNPT)
Female national pipe thread (FNPT)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.