missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
£52.80 - £349.00
Specifications
Material (Membrane) | Nylon |
---|---|
Filtration Area | 400 cm2 |
For Use With (Application) | Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. |
Housing Material | Polypropylene (PP) |
Max. Pressure | 4.1 bar (60 psi) |
Product Code | Brand | Pore Size | Inlet Style | Outlet Style | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Inlet Style | Outlet Style | Sterility | Price | Quantity & Availability | ||
11369084
|
Cytiva
6708-3602 |
0.2 μm | 1/2 SB inlet | 1/2 SB outlet | Sterile |
£63.35
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11379084
|
Cytiva
6709-3602 |
0.2 μm | MNPT inlet | SB outlet | Sterile |
£52.80
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11399084
|
Cytiva
6705-3604 |
0.45 μm | SB inlet | SB outlet | Sterile |
£64.45
Each |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10527111
|
Cytiva
6705-3602 |
0.2 μm | SB inlet | SB outlet | Sterile |
£112.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11329094
|
Cytiva
6706-3602 |
0.2 μm | SB inlet | SB outlet plus filling bell | Sterile |
£65.35
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11339094
|
Cytiva
2606T |
0.2 μm | FNPT | FNPT | Non-sterile |
£349.00
Pack of 5 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.- First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
- Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
- Housing thermally fused (no glues, adhesives, or extraneous materials).
- Integrity-testable by bubble point, pressure decay, or forward flow methods.
- Provides highly effective filtration area in a small size.
- Autoclavable; some presterilized with gamma irradiation.
- Manufactured in clean room facilities under ISO Quality Systems.
Specifications
Nylon | |
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
4.1 bar (60 psi) |
400 cm2 | |
Polypropylene (PP) |