missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 75 AS
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Outlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 5
Description
- Sterile and nonpyrogenic
- Integrity testable
- With inlet vent
- Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
- Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material (Membrane) | Nylon |
| Pore Size | 0.2 μm |
| Sterility | Sterile |
| Filtration Area | 820 cm2 |
| For Use With (Application) | Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. |
| Housing Material | Polypropylene |
| Inlet Style | Stepped Barb |
| Outlet Style | Stepped Barb |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction