missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polydisc In-line Filter
For serum samples
Brand: Cytiva 6724-5002
Code : 75
Additional Details : Weight : 0.31000kg
Description
- Glass microfiber and polyethersulfone membranes
- Contains stack of filter media
- Radiation sterilized
Specifications
PES | |
Sterile | |
10/Pk. |
0.2 μm | |
500.0mL/min./cm sq. |