missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polydisc Inline Filters
Versatile, cost-effective filters designed for larger volume sample filtration
£127.00 - £485.04
Specifications
For Use With (Application) | Larger volume sample filtration |
---|---|
Housing Material | Polypropylene (PP) |
Inlet Connection (Metric) | Tubing Nozzle 6 to 10 mm i.d. hose |
Diameter (Metric) | 50 mm |
Product Code | Brand | Material (Membrane) | Pore Size | Type | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Pore Size | Type | Quantity | Price | Quantity & Availability | ||
11349194
|
Cytiva
6724-5045 |
PES | 0.45 μm | Inline Filter | Pk. of 10 |
£178.00
Pack of 10 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11324634
|
Cytiva
6720-5045 |
PTFE | 0.45 μm | Polydisc TF | Pk. of 10 |
£127.00
Pack of 10 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11314634
|
Cytiva
6720-5002 |
PTFE | 0.2 μm | Polydisc TF | Pk. of 10 |
£214.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11334634
|
Cytiva
6721-5010 |
Polypropylene | 1 μm | Polydisc TF | Pk. of 10 |
£175.00
Pack of 10 |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11359194
|
Cytiva
6724-5145 |
PES | 0.45 μm | Polydisc AS | Pk. of 50 |
£485.04
Pack of 50 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Versatile, cost-effective filters designed for larger volume sample filtration
- Filter up to 1L with one device
- Can be used in conjunction with a syringe or connected inline (6-10mm I.D. hose) via stepped hose barbs
- High-purity polypropylene housing maintains sample purity
- 50mm filter devices
- Autoclavable
- Lightweight (11.5g)
- For tissue culture media, reagent preparation, particle counting solutions, pharmaceutical preparations High-throughput polyethersulfone membrane (low protein binding and no surfactants)
- Glass microfiber prefilter extends membrane life and effectively filters heavily contaminated samples
- Sterility cap on outlet (optional)
- No EtO residuals Barbed hose connections fit multiple tubing sizes Integrity testable (bubble point method) Sealed in its own “medical grade” clear blister pack and secured in a protective shelf pack
- Use in vents, inline, exhausts for growth environments, clean or sterile gases, solvents, reagents, drying gases, photoresists Hydrophobic PTFE membrane ideal for chemically aggressive solutions, reagents and organic solvents
- Chemical-resistant housing Integrity testable (bubble point or water breakthrough pressure in situ methods) Biosafe
Specifications
Larger volume sample filtration | |
Polypropylene (PP) | |
Tubing Nozzle 6 to 10 mm i.d. hose | |
50 mm |