missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polydisc TF In-line Filters

Versatile, reusable filters designed for large volume samples

Brand:  Cytiva 6720-5001

125.25 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : A0

Additional Details : Weight : 0.19000kg

Product Code. 11339204

  • £135.00 / Pack of 10
Estimated Shipment: 07-08-2024
to see stock.

Description

Description

Available options for filtration media suit a range of aqueous and organic samples

  • Hydrophobic PTFE membrane for filtration with chemically aggressive solutions, reagents and organic solvents
  • High-purity polypropylene housing maintains sample purity
  • 50mm filter
  • Can be used in conjunction with a syringe or connected in-line (6-10mm I.D. hose) via barbed hose connections
  • Autoclavable
  • Lightweight (11.5g)
  • Biosafe

  • 1μm device features polypropylene prefilter for use with heavy contamination

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

For in-line filtration and isolation applications in pharmaceuticals, biotech research, laboratories and electronics

Compliance

Integrity testable with bubble point method

Specifications

Specifications

Polydisc TF Filter, polypropylene, 0.1 μm (10 pcs)
0.1 μm
For sample preparation of ground water samples for the analysis of dissolved heavy metals
Tubing Nozzle 6 to 10 mm i.d. hose
PTFE
16 cm2
Polypropylene (PP)
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers