missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Circles

Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.

Brand:  Cytiva 1001-045

Code : A0

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 12494272

  • £4.61 / Pack of 100
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.

 

These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: >11μm
  • Porosity: medium
  • Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
  • Surface: smooth
  • Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
  • Used for soil analysis and seed testing procedures in the agricultural industry
  • Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
  • Atmospheric dust is collected from airflow and the stain-intensity measured 
  • Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection

 


Specifications

Specifications

13sec./100mL/sq. in.
Grade 1 Qualitative Filter Paper Standard Grade, circle, 14.5 cm
Cellulose
150 sec.
A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids.
Medium
180 μm
45 mm
87 g/m2
Disc
>11 μm
57mL/min. (water)
Grade 1
100/Pk.
Filter Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers