missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Sealable Hybridization Bags

Make hybridizations and incubations simpler

Brand:  Cytiva 10483064

109.26 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


Product Code. 11319374

  • £125.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Make hybridizations and incubations simpler

Use for hybridization and incubation reactions on thin wet membranes.

  • Open on three sides for easy insertion of wet membrane
  • Can be sealed with a standard heat-sealer
  • Size: 20.3 × 25.8cm (8 × 10 in.)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sealable Reaction Bag
Hybridization and incubation reaction
8 in.
8 in.
Heat Seal
50 Pack
20.3 cm
20.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.