missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Seed Testing Papers
Grade 181 seed testing circles
Brand: Cytiva 2181-090
20.35 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Grade 181 seed testing circles
Owing to their extremely high purity our seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances that could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.Specifications
Disc | |
Seed testing | |
Seed Test Paper | |
90 mm |
Cellulose | |
181 | |
3.54 in. |