missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ TurboBlotter Transfer System, Small Kits

Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA

Brand:  Cytiva 10416304

Code : 69

Additional Details : Weight : 1.13000kg

 View more versions of this product

Product Code. 10325561

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: Includes device plus five sets of transfer materials for gel size indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.

Description

Description

  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable

  • Model is 12 × 16cm, for gels from 7 × 8cm to 11 × 14cm

Order Info

Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).

Specifications

Specifications

DNA/RNA Transfer
Nytran™ SPC 0.45μm TurboBlotter™ Kit
5.5 in.
4.3 in.
11 x 14 cm Gel TurboBlotter™ Transfer System
12 x 16cm Small transfer device, 5 sets of transfer materials (set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o
14 cm
11 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers