Learn More
Cytiva Whatman™ Uniflo™ Syringe Filters With GF Pre-filter, 0.45 μm
Whatman™ Uniflo™ syringe filters are disposable filter units designed to provide clean filtrate from small volumes up to 100 mL.
Brand: Cytiva 9929-3004
Additional Details : Weight : 0.33000kg
Description
Whatman Uniflo GF Syringe Filters are available in a variety of membrane choices with a polypropylene overmold housing
- 0.2 μm or 0.45 μm pore sizes
- Laser etched printing on the filter for easy identification
- Bench top space saving packaging
Specifications
PTFE | |
0.45 μm | |
4.98 cm2 | |
Female Luer lock | |
100 Pk. | |
Glass Fiber (GF) |
Nylon | |
Non-sterile | |
Polypropylene | |
67.5 psi | |
Male Slip Luer | |
30 mm |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.