missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Uniflo™ Syringe Filters With GF Pre-filter, 0.45 μm

Whatman™ Uniflo™ syringe filters are disposable filter units designed to provide clean filtrate from small volumes up to 100 mL.

Brand:  Cytiva 9931-3004

Additional Details : Weight : 0.34000kg


 View more versions of this product

Product Code. 17345200

  • £265.00 / Pack of 100
Estimated Shipment: 12-12-2024
to see stock.
Add to basket

Description

Description

Whatman Uniflo GF Syringe Filters are available in a variety of membrane choices with a polypropylene overmold housing

  • 0.2 μm or 0.45 μm pore sizes
  • Laser etched printing on the filter for easy identification
  • Bench top space saving packaging
Specifications

Specifications

PTFE
0.45 μm
4.98 cm2
Female Luer lock
100 Pk.
Glass Fiber (GF)
H-PTFE
Non-sterile
Polypropylene
67.5 psi
Male Slip Luer
30 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FILTER GLASS CIRCLE

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers