missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UNIPLATE™ Microplates
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Brand: Cytiva 7701-5500
This item is not returnable.
View return policy
Description
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
- Withstand low temperatures (-80°C) for long-term storage
- Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
Specifications
Standard | |
6 x 8 | |
Polypropylene | |
48 | |
Flat Bottom (F-well) | |
25/Pk. |
UNIPLATE microplates | |
Natural | |
5 mL | |
Non-sterile | |
Ideal for homogeneous assay techniques utilized in high-throughput screening |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Whatman™ UNIPLATE™ Microplates > Flat well; Well volume: 5mL
Spot an opportunity for improvement?Share a Content Correction