missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Vacuum Filter Holders: Stainless Steel

Made from borosilicate glass with stainless steel support screen

Brand:  Cytiva 1960-032

Code : A0

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 11330644

  • £479.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Includes: Borosilicate glass funnel and base; anodized aluminum clamp; support screen (Type 304 stainless steel); rubber stopper (No. 5 for 25mm diameter filters to fit 125mL flask or No. 8 for all others to fit 1000mL flask).

Description

Description

Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.

  • Made from borosilicate glass
  • Ensure secure clamping and integral sealing at membrane edge
  • 47mm models have clearly printed graduation scales
  • For glass support screen, see cat. no. 099072

Applications:

  • Gravimetric analysis filtration
  • Ultraclean filtration
  • Bacteriological control of food, drink, pharmaceuticals, cosmetics
  • Removal of particulates in solution
  • Nucleic acid and protein analysis
  • HPLC solvent filtration

Specifications

Specifications

Membrane Holder
Borosilicate Glass
2.1 cm2
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers