Learn More
Cytiva Whatman™ ML050/0/03 Stainless Steel Frit for use with the ML 050 Stainless Steel Filter Holder for In-Line Filters
The Whatman™ ML050/0/03 Stainless Steel Frit is designed for use in in-line filters with the ML 050 Stainless Steel Filter Holder. Ideal for automated systems and high-speed batch filtration with robots, the steel build reduces contamination risk from radioactive/biohazardous materials.
Brand: Cytiva 10464103
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Product Type: Frit
- Model: ML050/0/03
- Material: Stainless Steel
- Includes: Ring
- For Use With (Application): Vacuum Filtration (In-Line Filters)
- For Use With (Equipment): Whatman™ ML 050 Stainless Steel Filter Holder
Specifications
ML 050 Stainless Steel Filter Holder | |
Vacuum Filtration (In-Line Filters) | |
ML050/0/03 |
Stainless Steel | |
Ring | |
Frit |