Learn More
Abnova™ Wheat Germ agglutinin native Protein from wheat germ
Used for Enzyme, SDS-PAGE
Brand:  Abnova™ P5361.5mg
Product Code. 16212280
  • £328.00 / 5mg
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Lyophilized from 0.05M Tris-HCl, 0.15M NaCl, pH 8.6
RUO
Lyophilized
≥94%
Agglutinin (Wheat Germ) Native Protein
5 mg
Store at -20°C. After reconstitution with deionized water containing 0.9% sodium chloride, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Enzyme Activity, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Plants