Learn More
Novus Biologicals™ Wheat Germ Agglutinin (WGA), Fluorescein
Brand:  Novus Biologicals™ FL-1021-10-NB
Product Code. 18391518
  • £385.00 / 10mg
Explore more special offers
Specifications
Wheat Germ Agglutinin (WGA), Fluorescein
10 mg
10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM CaCl2
5 mg/mL
Store at 4°C.