missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 12 x 32mm Polypropylene Vials with Limited Volume Insert

Limited Volume

Brand:  DWK Life Sciences 225180

 View more versions of this product

Product Code. 12634793

  • £302.00 / Pack of 1000
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 0.30mL volume
  • Offer good chemical resistance
  • Conical interior eliminates time-consuming assembly of vials and limited volume inserts
  • Designed for maximum retrieval of contents

TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 mm
0.3 mL
1.25 in.
Crimp Cap
1000 Cs.
Polypropylene
For maximum retrieval of contents
Limited volume insert
32 mm
11 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers