missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 250mL Open-Ended Flask
With 20 to 400 threads
Brand: DWK Life Sciences 297814
This item is not returnable.
View return policy
Description
- 20-400 top joint
- 20-400 bottom joint
Specifications
8.4 oz. | |
Open-ended Flask | |
250 mL |
Concentration of trace amounts of analytes in an organic solvent prior to analysis | |
20-400 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction