Learn More
DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert
For use with Wheaton CryoELITE™ Freestanding Cryogenic Vials
Brand:  DWK Life Sciences W224621-0303
63.89 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 12379959
  • £77.20 / Pack of 100
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
Description
Description
  • Makes sample identification and traceability reliable and convenient
  • Bar code insert manually press fits into bottom of vial
  • Data matrix readable with standard scanning hardware
  • Made from same cryogenic radiation-grade, virgin polypropylene as vial, ensuring uniform thermal expansion
Specifications
Specifications
Polypropylene
Sterile
500/Cs.