Learn More
DWK Life Sciences Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
Brand:  DWK Life Sciences 900205
 View more versions of this product
Product Code. 10512003
  • £52.40 / Pack of 6
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
  • Replacement dish and cover
  • Approx. inside bottom dimensions: 91 x 71 x 60mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Soda Lime Glass
Slide Staining Rack Handle
Glass Staining Dishes