Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
GREENER_CHOICE
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand:  DWK Life Sciences 224182-07
127.30 GBP valid until 2024-12-31
Use promo code "21972" to get your promotional price.
 View more versions of this product
Product Code. 11788779
  • £134.00 / Pack of 1000
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Green
1000/Cs.
13 mm
Aluminum
Center Disc Tear-out Cap