missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
GREENER_CHOICE

Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.

Brand:  DWK Life Sciences 224182-01

97.21 GBP valid until 2025-12-31
Use promo code "24362" to get your promotional price.


 View more versions of this product

Product Code. 10022372

  • £110.00 / Pack of 1000
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
1000/Cs.
13 mm
Aluminum
Center Disc Tear-out Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined > One-piece; Mouth O.D.: 13mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.