Learn More
DWK Life Sciences Wheaton™ BOD Bottles
Available with either glass robotic or pennyhead stoppers
£448.00 - £1530.00
Specifications
For Use With (Application) | Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action |
---|---|
Marking Spot | Yes |
Material | Borosilicate Glass |
Product Code | Brand | Barcode | Volume (Metric) | No. Sequence | Stopper | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Barcode | Volume (Metric) | No. Sequence | Stopper | Price | Quantity & Availability | ||
12473711
|
DWK Life Sciences
227494-99 |
Yes | 60 mL | Specials | Robotic |
£1530.00
Pack of 36 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12463741
|
DWK Life Sciences
227497-13G |
Yes | 300 mL | 289 to 312 | Pennyhead |
£483.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11718879
|
DWK Life Sciences
227497-03G |
Yes | 300 mL | 49 to 72 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
10678714
|
DWK Life Sciences
227494-00 |
No | 60 mL | Unnumbered | Robotic |
£1155.00
Pack of 36 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12433711
|
DWK Life Sciences
227494-02 |
Yes | 60 mL | 37 to 72 | Pennyhead |
£1375.00
Pack of 36 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11764179
|
DWK Life Sciences
227494-01 |
Yes | 60 mL | 1 to 36 | Robotic |
£1375.00
Pack of 36 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11728879
|
DWK Life Sciences
227497-04G |
Yes | 300 mL | 73 to 96 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11678308
|
DWK Life Sciences
227497-00 |
Yes | 300 mL | Unnumbered | Robotic |
£448.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
10590463
|
DWK Life Sciences
227497-01 |
Yes | 300 mL | 1 to 24 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12731638
|
DWK Life Sciences
227497-02 |
Yes | 300 mL | 25 to 48 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12453721
|
DWK Life Sciences
227497-03 |
Yes | 300 mL | 49 to 72 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12473721
|
DWK Life Sciences
227497-04 |
Yes | 300 mL | 73 to 96 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12413731
|
DWK Life Sciences
227497-06 |
Yes | 300 mL | 121 to 144 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12493721
|
DWK Life Sciences
227497-05 |
Yes | 300 mL | 97 to 120 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12433731
|
DWK Life Sciences
227497-07 |
Yes | 300 mL | 145 to 168 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12453731
|
DWK Life Sciences
227497-08 |
Yes | 300 mL | 169 to 192 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12473731
|
DWK Life Sciences
227497-09 |
Yes | 300 mL | 193 to 216 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12493731
|
DWK Life Sciences
227497-10 |
Yes | 300 mL | 217 to 240 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12413741
|
DWK Life Sciences
227497-11 |
Yes | 300 mL | 241 to 264 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12453741
|
DWK Life Sciences
227497-13 |
Yes | 300 mL | 289 to 312 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12433741
|
DWK Life Sciences
227497-12 |
Yes | 300 mL | 265 to 288 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12473741
|
DWK Life Sciences
227497-14 |
Yes | 300 mL | 313 to 336 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12493741
|
DWK Life Sciences
227497-15 |
Yes | 300 mL | 337 to 360 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12413751
|
DWK Life Sciences
227497-16 |
Yes | 300 mL | 361 to 384 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12433751
|
DWK Life Sciences
227497-17 |
Yes | 300 mL | 385 to 408 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12453751
|
DWK Life Sciences
227497-18 |
Yes | 300 mL | 409 to 432 | Robotic |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11734189
|
DWK Life Sciences
227497-01G |
Yes | 300 mL | 1 to 24 | Pennyhead |
£483.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12403721
|
DWK Life Sciences
227497-00G |
Yes | 300 mL | Unnumbered | Pennyhead |
£460.00
Pack of 24 |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
11764219
|
DWK Life Sciences
227497-02G |
Yes | 300 mL | 25 to 48 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12403731
|
DWK Life Sciences
227497-05G |
Yes | 300 mL | 97 to 120 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12423731
|
DWK Life Sciences
227497-06G |
Yes | 300 mL | 121 to 144 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12443731
|
DWK Life Sciences
227497-07G |
Yes | 300 mL | 145 to 168 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12463731
|
DWK Life Sciences
227497-08G |
Yes | 300 mL | 169 to 192 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12483731
|
DWK Life Sciences
227497-09G |
Yes | 300 mL | 193 to 216 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12403741
|
DWK Life Sciences
227497-10G |
Yes | 300 mL | 217 to 240 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
12423741
|
DWK Life Sciences
227497-11G |
Yes | 300 mL | 241 to 264 | Pennyhead |
£473.00
Pack of 24 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
Description
Available with either glass robotic or pennyhead stoppers
Bottles
- Borosilicate glass, 60 or 300mL, numbered or unnumbered
- With permanent marking areas; numbered versions are also bar-coded
- Specially designed shoulder radius sweeps all air from inside the bottle during filling
- 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
- Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
- Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
- Stopper joint is compatible with the probes of the leading BOD and DO meters
- Glass robotic stoppers are compatible with most robotic systems
Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.
Order Info
Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)
Specifications
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action | |
Yes | |
Borosilicate Glass |