Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Use as replacement caps or for vials that are provided without caps
Brand: DWK Life Sciences 241017
Additional Details : Weight : 3.62870kg
Description
Screw Caps For Liquid Scintillation Vials
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
Specifications
Cap Size: 22-400 | |
White | |
Screw Cap | |
Liquid Scintillation Vial |
Metal Foil | |
Urea | |
22-400 |