missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket

Provide maximum surface between culture and glass flask

Brand:  DWK Life Sciences 356884

511.85 GBP valid until 2024-12-31
Use promo code "21939" to get your promotional price.


 View more versions of this product

Product Code. 10374601

  • £694.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provide maximum surface between culture and glass flask

  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silicone Rubber
1000 mL
PTFE Resin
10 to 44 cm
4.5 cm
Flask, impeller and caps
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers