missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps Product Code.: 10021502
Estimated Shipment: 13-03-2025
to see stock.
Add to basket

DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps

Product Code. 10021502 Shop All Wheaton Science Products Products
250 mL, Convenience pack
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk pack
This item is not returnable. View return policy

Product Code. 10021502

Brand: DWK Life Sciences W216803

Estimated Shipment: 13-03-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for solvent, chemical and sample storage

  • Narrow mouth enables liquids to be carefully poured to prevent spillage
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

  • Available in partitioned Convenience or Valu-Bulk™ cartons

Order Info

Caps can be ordered separately.

Compliance

USP Type-III soda-lime glass

TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Clear
Diameter (English) 2.36 in.
Material Soda Lime Glass
Packaging Convenience pack
Closure Color White
Certifications/Compliance EPA, FDA
Height (Metric) 138 mm
Height (English) 5.43 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 24-400 mm
Diameter (Metric) 60 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.