missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached

Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.

Brand:  DWK Life Sciences W224581

336.30 GBP valid until 2024-12-31
Use promo code "21960" to get your promotional price.


 View more versions of this product

Product Code. 10376942

  • £375.00 / Pack of 288
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.

  • Offer a variety of uses ranging from sample storage to freeze-drying
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Cap and Cap Liners
    • Solid top, black phenolic caps
    • 14B rubber-lined black phenolic screw cap for aqueous samples
    • PTFE/14B rubber-lined black phenolic screw cap for organic samples
  •  

Order Info

Replacements caps may be purchased separately.

Compliance

Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE/14B Rubber
12 mm
2 mL
Screw
Wheaton
Phenolic
Screw Cap
ASTM Type I, Class A, USP Type I
8-425
Clear
Borosilicate Glass (Low Extractable)
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
0.5 dr.
2 mL
Autoclavable
38 mm
Vial
M-T File™ and Vial Rack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.