Learn More
DWK Life Sciences Wheaton™ Clear Standard Wide-Mouth Bottles: With Polyvinyl-Lined Caps
Ideal for the storage of dry materials and specimens
£34.20 - £70.15
Specifications
Color Clear
Material Soda Lime Glass
Closure Color White
Closure Liner Poly/Vinyl
Closure Material Polypropylene
View More Specs
Products 5
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12359829
View Documents
DWK Life Sciences
W216924
125 mL
£37.75
Pack of 24
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
12369829
View Documents
DWK Life Sciences
W216925
250 mL
£47.10
Pack of 24
Estimated Shipment: 29-07-2025
to see stock.
Add to basket
12379829
View Documents
DWK Life Sciences
W216926
500 mL
£70.15
Pack of 24
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
12389829
View Documents
DWK Life Sciences
W216927
2000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12399829
View Documents
DWK Life Sciences
W216928
4000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Clear for easy viewing of contents
  • Wide mouth for easy access to contents
  • Rounded shoulders
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear Type III soda-lime glass
Specifications
Specifications
Clear
White
Polypropylene
Soda Lime Glass
Poly/Vinyl
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Clear Standard Wide-Mouth Bottles: With Polyvinyl-Lined Caps