missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars without Caps
24.52 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Use promo code "23261" to get your promotional price.
Click to view available options
Capacity:
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Diameter (English):
2.16 in.
2.36 in.
2.8 in.
3.5 in.
3.7 in.
Height (English):
1.8 in.
2.6 in.
3.4 in.
3.7 in.
6.6 in.
Description
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
- Chemical-resistant caps conform to EPA and FDA regulations
- Available in partitioned Convenience or Valu-Bulk™ cartons
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
- Evident labels to protect sample integrity in transit or long-term storage
Specifications
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Clear |
Diameter (English) | 3.7 in. |
Material | Soda Lime Glass |
Height (Metric) | 170 mm |
Height (English) | 6.6 in. |
Closure Size | 89-400 mm |
Diameter (Metric) | 95 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'