missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits

Product Code. 17962571 Shop All Wheaton Science Products Products
Click to view available options
Closure Color:
Blue
Red
Closure Type:
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure Serum Stopper
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17962571

Brand: DWK Life Sciences W991015CPA

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Red
Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Volume (Metric) 10 mL
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Graduated No
Includes Vial, Stopper, and Seal
No. per Case 104/Cs.
Style Crimp top
Product Line Wheaton CompletePAK
Closure Type Ultra Pure Serum Stopper
Height (Metric) 50 mm
Closure Size 20 mm
Marking Spot No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.