missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

£1335.00 - £3210.00

Specifications

Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
View More Specs

Products 18
Product Code Brand Closure Color Closure Type Volume (Metric) Price Quantity & Availability  
Product Code Brand Closure Color Closure Type Volume (Metric) Price Quantity & Availability  
17912641
View Documents
DWK Life Sciences
W991002-CPA
Blue OmniFlex 3G Igloo Lyo Stopper 2 mL
£3210.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17952641
View Documents
DWK Life Sciences
W991005-CPA
Red OmniFlex 3G Igloo Lyo Stopper 2 mL
£3205.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17932581
View Documents
DWK Life Sciences
W991001-CPA
Blue OmniFlex 3G Serum Stopper 2 mL
£3055.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17962641
View Documents
DWK Life Sciences
W991004-CPA
Red OmniFlex 3G Serum Stopper 2 mL
£3060.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17922641
View Documents
DWK Life Sciences
W991000-CPA
Blue Ultra Pure Serum Stopper 2 mL
£2795.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17932641
View Documents
DWK Life Sciences
W991003-CPA
Red Ultra Pure Serum Stopper 2 mL
£2795.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17932591
View Documents
DWK Life Sciences
W991008-CPA
Blue OmniFlex 3G Igloo Lyo Stopper 5 mL
£1490.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17912651
View Documents
DWK Life Sciences
W991011-CPA
Red OmniFlex 3G Igloo Lyo Stopper 5 mL
£1490.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17982641
View Documents
DWK Life Sciences
W991007-CPA
Blue OmniFlex 3G Serum Stopper 5 mL
£1480.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17992641
View Documents
DWK Life Sciences
W991010-CPA
Red OmniFlex 3G Serum Stopper 5 mL
£1450.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17992581
View Documents
DWK Life Sciences
W991006-CPA
Blue Ultra Pure Serum Stopper 5 mL
£1335.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17972641
View Documents
DWK Life Sciences
W991009-CPA
Red Ultra Pure Serum Stopper 5 mL
£1335.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17902651
View Documents
DWK Life Sciences
W991014-CPA
Blue OmniFlex 3G Igloo Lyo Stopper 10 mL
£1740.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17942651
View Documents
DWK Life Sciences
W991017-CPA
Red OmniFlex 3G Igloo Lyo Stopper 10 mL
£1710.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17962591
View Documents
DWK Life Sciences
W991013-CPA
Blue OmniFlex 3G Serum Stopper 10 mL
£1700.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17932651
View Documents
DWK Life Sciences
W991016-CPA
Red OmniFlex 3G Serum Stopper 10 mL
£1660.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17922651
View Documents
DWK Life Sciences
W991012-CPA
Blue Ultra Pure Serum Stopper 10 mL
£1560.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17962571
View Documents
DWK Life Sciences
W991015-CPA
Red Ultra Pure Serum Stopper 10 mL
£1560.00
1 set
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
Description

Description

  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions
Specifications

Specifications

Amber
No
Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Vial, Stopper, and Seal
Wheaton CompletePAK
Borosilicate Glass
Sterile
No
Crimp top
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.