missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits
Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
Omni Flex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Sterile
OmniFlex 3G Sterile Igloo Lyo Stopper
OmniFlex 3G Sterile Serum Stopper
Ultra Pure Sterile
Ultra Pure Sterile Serum Stopper
Color (Seal):
Blue
Red
Unit Size:
Pack of 158
Pack of 200
Pack of 210
Description
Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Kitted solutions feature equal amounts of RtU components for reduced scrap rates
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- CompletePAK™ ModPAK Bulk configurations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
| Color (Seal) | Blue |
| Color | Clear |
| Sterility | Sterile |
| Volume (Metric) | 2 mL |
| Includes | Vial, Stopper, and Seal |
| No. per Case | 210 |
| Product Line | Wheaton CompletePAK |
| Closure Type | OmniFlex 3G Sterile |
| Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
| Closure Size | 13 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction