missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit

Product Code. 17912671 Shop All Wheaton Science Products Products
Click to view available options
Closure Color:
Blue
Red
Closure Type:
Omni Flex Igloo Lyo Stopper
Omni Flex Serum Stopper
Ultra Pure Serum Stopper
Quantity:
104/Pk.
278/Pk.
missing translation for 'unitSize'
1 set
Diameter (Metric) Outer:
22 mm
24 mm
This item is not returnable. View return policy

Product Code. 17912671

missing translation for 'mfr': DWK Life Sciences W991005ACPMP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Six inventory matched kits conveniently packed into a single shipper case.
  • Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Focuses your effort on core competencies and reduces lead time
  • Streamlines process and frees up valuable clean room space USP /EP 2.9.19
  • Particulate Matter in Injections, USP /EP 2.6.1,
  • The Bacterial Endotoxins Test and USP / EP 2.6.14
  • The Sterility Test, USP /EP 3.2.1
  • Glass Containers for Pharmaceutical Use
  • USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time
  • Minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in amber for photosensitive contents
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Red
Color Amber
Material Borosilicate Glass
Sterility Sterile
Disposable Single-use
For Use With (Application) Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging
Graduated No
Includes Vial, Stopper, and Seal
Product Line CompletePAK ModPAK
Closure Material Bromobutyl
Autoclavable Autoclavable
Closure Type Omni Flex Igloo Lyo Stopper
Height (Metric) 32 mm
Quantity 278/Pk.
Certifications/Compliance USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Type Serum Vial
Closure Size 13 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.