Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand: DWK Life Sciences W986002-S
This item is not returnable.
View return policy
Description
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Amber versions for light sensitive content packaging
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Clear | |
Sterile | |
210 | |
USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
Borosilicate Glass (Type 1) | |
10 mL | |
Wheaton CompletePAK |
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials > 10 mL; Clear
Spot an opportunity for improvement?Share a Content Correction