missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps

The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.

Brand:  DWK Life Sciences W125495

157.41 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12477371

  • £202.00 / Pack of 6
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Connector
13-425, 22-400
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.