Learn More
DWK Life Sciences Wheaton™ Crimpers
With ergonomically designed handles for sealing standard laboratory vials
£481.00 - £570.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
11591215
|
DWK Life Sciences
W225300 |
8 mm Seals |
£570.00
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
10495951
|
DWK Life Sciences
W225302 |
13 mm Seals |
£488.00
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
10670884
|
DWK Life Sciences
W225301 |
11 mm Seals |
£481.00
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
10432722
|
DWK Life Sciences
W225303 |
20 mm Seals |
£488.00
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
Description
- Wheaton E-Z Crimpers feature cushioned ergonomic handles that aid in reducing hand fatigue and add a high degree of comfort for the user. Each crimper size is labeled for quick identification. Other features include polished crimping jaws for consistent sealing, a short handle and redesigned spring assembly that further aids in reducing hand fatigue. For semi-automatic crimping, use the E-Z Crimper with the Wheaton Crimpmaster™ Crimping Station. An adjustable stop allows you to determine the tightness of the seal while maintaining consistent crimping. Tapered jaws provide for easier vial access when using racks. The 11 mm E- Z Crimper is supplied with a plunger insert to give the option of crimping the top of a vial creating an extra tight seal. We recommend this only for standard opening vials.
- Crimpers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Warranty
Two years
Specifications
Case of 1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Crimpers