missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ CryoFILE™ Tissue Vial Storage Boxes

Storage Boxes for batching and storing Wheaton CryoELITE Tissue Vials (Cat. No. 13-709-140)

Brand:  DWK Life Sciences W651610-B

 View more versions of this product

Product Code. 15500326

  • £124.00 / Pack of 15
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CryoFILE partitions are numbered from 1-25 for easy content identification, with the numbering system printed on lid and bottom of box. Available in six colors, to provide easy sample identification. Made of water-repellent material for longer durability, the CryoFILE Tissue Storage Boxes are able to withstand cryogenic temperatures to -196°C.

  • Holds 25 tissue vials
  • Used for batching and storing cryogenic tissue vials
  • Two boxes can be stacked in a standard 2 in. freezer rack, allowing for storage of 50 vials in standard racks
  • Partitions numbered 1-25 for easy content identification
  • Water repellent material allows for longer durability
  • Dimensions: 130 x 130 x 25mm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cryo Storage Box
25
25 mm
-384.8°F
Square
Blue
50 Vials
130 mm
-196°C
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers