Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
Brand: DWK Life Sciences 281155
This item is not returnable.
View return policy
Description
- Utilizes clear-seal grease-free joints
- «O« ring not required
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
20 cm | |
Diazomethane Generator |
7.8 in. | |
O-ring clamp, a 13-425 screw cap with a cut-out top and a ptfe faced rubber liner |