missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Disposable Drum Thieves

Economical and convenient to use

Brand:  DWK Life Sciences 885300

 View more versions of this product

Product Code. 12497311

  • £309.00 / Pack of 100
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simply insert empty tube into drum, hold thumb over the top, and withdraw sample
  • Soda-lime glass units break easily for quick disposal
  • Length: 43 in. (109cm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Drum Thief
25 mL
Ideal for quick and easy sample retrieval
0.75 cm
109.2 cm
Soda Lime Glass
Single-use
0.29 in.
43 in.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Sonderangebote

Sonderangebote