missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Disposable Glass Coliwasa Sampler

Constructed of soda-lime glass with liquid-tight PTFE-lined seal

Brand:  DWK Life Sciences 885220

Product Code. 12477301

  • £234.00 / Pack of 12
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prescored components allow for easy disposal
  • PTFE seal locks the sampled column in place
  • Transfer sample to appropriate container without any loss

Order Info

Dimensions: 0.88 in. dia. x 42 in.L (2.2 x 107cm); Tapered bottom with 0.5 in. dia. (1.3cm) opening

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coliwasa Sampler
PTFE
200 mL
0.5 in.
1.3 cm
41.92 in.
Soda Lime Glass
6.76 oz.
Single-use
0.86 in.
2.2 cm
106.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.