Learn More
DWK Life Sciences Wheaton™ E-Z Crimpers
Attach seals easily with these high-quality crimpers
£658.00 - £798.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10712573
|
DWK Life Sciences
224307 |
30 mm Aluminum Seals |
£798.00
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
11788879
|
DWK Life Sciences
224322 |
13 mm Flip-cap Seals |
£658.00
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
10276284
|
DWK Life Sciences
224323 |
20 mm Flip-cap Seals |
£684.00
Each |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
Description
- Labeled for quick identification
- Cushioned, short ergonomic handle
- Polished crimping jaws
- Spring assembly
- Adjustable stop
Specifications
Not Autoclavable |