missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ E-Z Decappers

Product Code. 11511225 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
11 mm Seals
13 mm Seals
20 mm Seals
8 mm Seals
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11511225

Brand: DWK Life Sciences W225350

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

With ergonomically designed handles for sealing and decapping standard laboratory vials

Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.

  • Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
  • The decappers come with visible size labels for easy identification.
  • User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
  • Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C

Warranty

Two years

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 8 mm Seals
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.