missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ E-Z Vials™

Glass autosampler vials featuring a large opening

Brand:  DWK Life Sciences 225179

 View more versions of this product

Product Code. 10614343

  • £227.00 / Pack of 1000
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 12mm dia. × 32mm H (0.47 × 1.26 in.)
  • 11mm (0.43 in.) crimp top
  • 1.8mL (0.06oz)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Silica Glass
1.25 in.
32 mm
11 mm
12 mm
1.8 mL
Crimp/Snap
100 Pk.
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers