Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
£96.55 - £336.00
Specifications
Material | Borosilicate Glass |
---|---|
Thread Style | Screw |
Autoclavable | Autoclavable |
Closure Type | Screw Cap |
For Use With (Equipment) | M-T File™ and Vial Rack |
Product Code | Brand | Volume (Metric) | Certifications/Compliance | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Certifications/Compliance | Color | Price | Quantity & Availability | |||
10380675
|
DWK Life Sciences
224833 |
40 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£298.00
Pack of 200 |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||
10792477
|
DWK Life Sciences
224836 |
40 mL | USP Type I | Amber |
£336.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
11541195
|
DWK Life Sciences
224831 |
20 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£262.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10411485
|
DWK Life Sciences
224808 |
24 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£249.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10584805
|
DWK Life Sciences
224806 |
16 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£184.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10330395
|
DWK Life Sciences
224801 |
2 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£124.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10160671
|
DWK Life Sciences
224804 |
8 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£153.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10400743
|
DWK Life Sciences
224805 |
12 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£178.00
Pack of 200 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||
11738929
|
DWK Life Sciences
224812 |
4 mL | USP Type I | Amber |
£150.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
13171803
|
DWK Life Sciences
224814 |
8 mL | USP Type I | Amber |
£176.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10218844
|
DWK Life Sciences
224832 |
25 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£307.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10567581
|
DWK Life Sciences
224802 |
4 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£131.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
10399043
|
DWK Life Sciences
224704 |
8.0 mL | ASTM Type I, Class A, USP Type I | Clear |
£96.55
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
12464791
|
DWK Life Sciences
W224815 |
20 mL | USP Type I | Amber |
£256.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
12474791
|
DWK Life Sciences
W224834 |
30 mL | ASTM E438, USP Type I, Type I, Class A | Clear |
£290.00
Pack of 200 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
- Autoclavable
- Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
- Amber vials made from borosilicate glass that protects light-sensitive products
Order Info
Caps available separately
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
Specifications
Borosilicate Glass | |
Autoclavable | |
M-T File™ and Vial Rack |
Screw | |
Screw Cap |