missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Glass Serum Bottles
Shop All Wheaton Science Products Products
Click to view available options
250mL
Description
- Raised graduations indicate approximate capacities in milliliters
- Accepts standard stoppers and seals
Specifications
Specifications
Capacity | 8.45 oz., 250mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Color | Clear |
Diameter (English) | 2.36 in. |
Diameter (Metric) Outer Neck | 30mm |
Diameter (Metric) Inner Neck | 15mm |
Material | Borosilicate Glass |
Height (Metric) | 158 mm |
Product Type | Serum Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction