missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Glass Serum Bottles
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
500 mL
Description
WHEATON™ Glass Serum Bottles are ideal for long term sample storage and drug containers.
Specifications
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Clear |
Diameter (English) | 2.95 in. |
Material | Borosilicate Glass |
Height (Metric) | 190 mm |
Product Type | Serum Bottle |
Height (English) | 7.48 in. |
Diameter (Metric) | 75 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction