Learn More
DWK Life Sciences Wheaton™ Glass Staining Dishes for 50 Slides
Holds up to 50 slides horizontally
Brand:  DWK Life Sciences 900402
 View more versions of this product
Product Code. 10254732
  • £204.00 / Pack of 3
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dish and cover are made from soda-lime glass
  • Dimensions: 3.25 L × 7.875 W × 4.125 in. H (80 × 200 × 105 mm)
Rack
  • Nontarnishable and withstands most staining solutions
  • Holds up to 50 slides horizontally
  • Hinged handle allows easy insertion and removal of slides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
Cover
Cover
Soda Lime Glass
Glass staining dish for 50 slides